Procain Hydrochlorid 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride

A17485.18 A17485.30
A17485.18EA 31.8 EUR
A17485.18 A17485.30
Procain Hydrochlorid 99%
Procain Hydrochlorid

Purity: 99% CAS No.: 51-05-8 Molecular Formula: C13H20N2O2.HCl Formula Weight: 272.78 Odour: Odourless Melting Point: 155-157° Boiling Point: 195-196°/17mm Merck Reference: 14,7757 UN No.: 2811 MDL No.: MFCD00013000

Formel: C₁₃H₂₁ClN₂O₂
Molecular Weight: 272,77 g/mol
Siedepunkt: 195…196 °C (17 mmHg)
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013000
CAS-Nummer: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7757

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR