Procain Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

P9879-100G P9879-250G P9879-50G
SIALP9879-100GEA 65.5 EUR
SIALP9879-100G SIALP9879-250G SIALP9879-50G
Procain Hydrochlorid, Sigma-Aldrich®
Procain Hydrochlorid
Formel: C₁₃H₂₁ClN₂O₂
MDL: MFCD00013000
CAS-Nummer: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR