Rhodanin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one
A18293.14
A18293.22
A18293.36
A18293.14EA
50
EUR
A18293.14
A18293.22
A18293.36
Rhodanin ≥98%
Rhodanin
Purity: 98+%
CAS No.: 141-84-4
Molecular Formula: C3H3NOS2
Formula Weight: 133.19
Odour: Odourless
Melting Point: 166-171°
Beilstein Reference: 110701
Merck Reference: 14,8184
MDL No.: MFCD00005488
Formel:
C₃H₃NOS₂ Molecular Weight: 133,19 g/mol Schmelzpunkt: 166…171 °C Dichte: 0,868 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005488 CAS-Nummer: 141-84-4 EINECS: 205-505-1 Merck Index: 14,8184 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...