Rhodanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

M-Clarity™ Qualitätsstufe = MQ100

118192-100G 118192-25G
SIAL118192-100GEA 104 EUR
SIAL118192-100G SIAL118192-25G
Rhodanin, Sigma-Aldrich®
Rhodanin
Formel: C₃H₃NOS₂
Molecular Weight: 133,19 g/mol
MDL: MFCD00005488
CAS-Nummer: 141-84-4
EINECS: 205-505-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR