2,4-Thiazolidindion ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,3-Thiazolidine-2,4-dione
A13496.14
A13496.22
A13496.14EA
62.8
EUR
A13496.14
A13496.22
2,4-Thiazolidindion ≥99%
2,4-Thiazolidindion
Purity: 99%
CAS No.: 2295-31-0
Molecular Formula: C3H3NO2S
Formula Weight: 117.13
Melting Point: 123-126°
Boiling Point: 178-179°/19mm
MDL No.: MFCD00005478
Formel:
C₃H₃NO₂S Molecular Weight: 117,13 g/mol Siedepunkt: 178…179 °C (19 mmHg) Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005478 CAS-Nummer: 2295-31-0 EINECS: 218-941-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...