1,2-Phenylendiessigsäure 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

157550050 157550250
ACRO157550050EA 41.6 EUR
ACRO157550050 ACRO157550250
1,2-Phenylendiessigsäure 99%
1,2-Phenylendiessigsäure
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004329
CAS-Nummer: 7500-53-0
EINECS: 231-355-1

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 150 to 152 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR