1,4-Phenylendiessigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,2'-(1,4-Phenylene)diacetic acid
M-Clarity™ Qualitätsstufe = MQ100
P23407-25G
P23407-5G
SIALP23407-25GEA
257
EUR
SIALP23407-25G
SIALP23407-5G
1,4-Phenylendiessigsäure, Sigma-Aldrich®
1,4-Phenylendiessigsäure
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol |
MDL:
MFCD00004354 CAS-Nummer: 7325-46-4 EINECS: 230-801-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...