5-Brombenzo[b]thiophen ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromobenzothiophene
L20133.03
L20133.06
L20133.03EA
83.8
EUR
L20133.03
L20133.06
5-Brombenzo[b]thiophen ≥98%
5-Brombenzo[b]thiophen
Purity: 98+%
CAS No.: 4923-87-9
Molecular Formula: C8H5BrS
Formula Weight: 213.10
Melting Point: 44-48°
Boiling Point: 105-107°/10mm
MDL No.: MFCD03069318
Formel:
C₈H₅BrS Molecular Weight: 213.1 g/mol Siedepunkt: 105…107 °C (10 mmHg) Schmelzpunkt: 44…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03069318 CAS-Nummer: 4923-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...