Thiophen-2-carbonsäurechlorid 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

187770250 187771000
ACRO187770250EA 34.5 EUR
ACRO187770250 ACRO187771000
Thiophen-2-carbonsäurechlorid 98%
Thiophen-2-carbonsäurechlorid
Formel: C₅H₃ClOS
Molecular Weight: 146,6 g/mol
Dichte: 1,37 g/cm³
Flammpunkt: 90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005428
CAS-Nummer: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to green Liquid
Infrared spectrum Conforms
Refractive index 1.5890 to 1.5910 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR