Thiophen-2-carbonsäurechlorid

Lieferant: Thermo Scientific

Synonyme: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

CC13002.250 CC13002.1
MAYBCC13002.250EA 0 EUR
MAYBCC13002.250 MAYBCC13002.1
Thiophen-2-carbonsäurechlorid
Thiophen-2-carbonsäurechlorid
Formel: C₅H₃ClOS
Molecular Weight: 146.6 g/mol
MDL: MFCD00005428
CAS-Nummer: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR