3-Methylthiophen-2-carbaldehyd

Lieferant: Thermo Scientific

Synonyme: 3-Methyl-2-thiophenecarboxaldehyde, 3-Methylthiophene-2-carbaldehyde

SB00927.1 SB00927.10 SB00927.50
MAYBSB00927.1EA 0 EUR
MAYBSB00927.1 MAYBSB00927.10 MAYBSB00927.50
3-Methylthiophen-2-carbaldehyd
3-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
Molecular Weight: 126,18 g/mol
MDL: MFCD00005430
CAS-Nummer: 5834-16-2
EINECS: 227-418-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR