3-Methylthiophen-2-carbaldehyd
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methyl-2-thiophenecarboxaldehyde, 3-Methylthiophene-2-carbaldehyde
HI-1253-5G
HI-1253-25G
HI-1253-100G
COBBHI-1253-5GEA
35.7
EUR
COBBHI-1253-5G
COBBHI-1253-25G
COBBHI-1253-100G
3-Methylthiophen-2-carbaldehyd
3-Methylthiophen-2-carbaldehyd
Formel:
C₆H₆OS Molecular Weight: 126,18 g/mol |
MDL:
MFCD00005430 CAS-Nummer: 5834-16-2 EINECS: 227-418-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...