(R)-(-)-2-Oxothiazolidin-4-carbonsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: (R)-2-Oxothiazolidine-4-carboxylic acid, Procysteine

4012415.0001 4012415.0005
BACE4012415.0001EA 0 EUR
BACE4012415.0001 BACE4012415.0005
(R)-(-)-2-Oxothiazolidin-4-carbonsäure
(R)-(-)-2-Oxothiazolidin-4-carbonsäure
Formel: C₄H₅NO₃S
Molecular Weight: 147.15 g/mol
MDL: MFCD00066092
CAS-Nummer: 19771-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR