(R)-(-)-Thiazolidin-4-carbonsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid
138640250
138641000
138640100
ACRO138640250EA
53.7
EUR
ACRO138640250
ACRO138641000
ACRO138640100
(R)-(-)-Thiazolidin-4-carbonsäure 98%
(R)-(-)-Thiazolidin-4-carbonsäure
Formel:
C₄H₇NO₂S Molecular Weight: 133,17 g/mol Schmelzpunkt: 203 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005212 CAS-Nummer: 34592-47-7 EINECS: 252-106-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Specific optical rotation | -136° to -144° (20°C, 589 nm) (c=1, H2O) |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...