(R)-(-)-Thiazolidin-4-carbonsäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid
A15033.09
A15033.18
A15033.30
A15033.09EA
31.2
EUR
A15033.09
A15033.18
A15033.30
(R)-(-)-Thiazolidin-4-carbonsäure ≥98%
(R)-(-)-Thiazolidin-4-carbonsäure
Purity: 98%
CAS No.: 34592-47-7
Molecular Formula: C4H7NO2S
Formula Weight: 133.17
Melting Point: 192-200° dec.
Beilstein Reference: 81065
Merck Reference: 14,9445
Optical Rotation: -141° (c=1.3 in water)
UN No.: 2811
MDL No.: MFCD00005212
Formel:
C₄H₇NO₂S Molecular Weight: 133,17 g/mol Schmelzpunkt: 192…200 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005212 CAS-Nummer: 34592-47-7 EINECS: 252-106-3 ADR: 6.1,III Merck Index: 14,9445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...