9-Fluorenylmethylchlorformiat (Fmoc-Chlorid), Supelco®

Lieferant: Merck

Synonyme: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

M-Clarity™ Qualitätsstufe = MQ100

23186-1G 23186-5G
23186-1GEA 54.9 EUR
23186-1G 23186-5G
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid), Supelco®
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
Formel: C₁₅H₁₁ClO₂
Molecular Weight: 258,7 g/mol
MDL: MFCD00001138
CAS-Nummer: 28920-43-6
EINECS: 249-313-6
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1da9ad65ec6920fa35b7911c09832b0c6e043d1d&vid=3c23525972015b27dd0f1f9d4c1c8aaa23e03d93&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR