4-Ethoxymethylen-2-phenyl-2-oxazolin-5-on ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: (4E)-4-(Ethoxymethylene)-2-phenyl-1,3-oxazol-5(4H)-one, 4-Ethoxymethylene-2-phenyloxazolin-5-one
A10155.03 A10155.06
A10155.03EA 38.1 EUR
A10155.03 A10155.06
4-Ethoxymethylen-2-phenyl-2-oxazolin-5-on ≥98%
4-Ethoxymethylen-2-phenyl-2-oxazolin-5-on
Purity: 98+% CAS No.: 15646-46-5 Molecular Formula: C12H11NO3 Formula Weight: 217.22 Storage Temperature: 2°C to 8°C Melting Point: 93-97° Beilstein Reference: 163055 MDL No.: MFCD00003204
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,22 g/mol
Schmelzpunkt: 93…97 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03225480
CAS-Nummer: 15646-46-5
EINECS: 239-713-9