2-Ethyl-2-oxazolin ≥99%

Lieferant: Thermo Scientific
Gefahr

B23287.22 B23287.36
B23287.22EA 50.2 EUR
B23287.22 B23287.36
2-Ethyl-2-oxazolin ≥99%
2-Ethyl-2-oxazolin

Purity: 99% CAS No.: 10431-98-8 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 29°(84°F) Refractive Index: 1.4370 Odour: Amine-like pH: 10-12 Melting Point: -62° Boiling Point: 128-129° Beilstein Reference: 105464 UN No.: 2734 MDL No.: MFCD00005302

Formel: C₅H₉NO
Molecular Weight: 99,13 g/mol
Siedepunkt: 128…129 °C
Schmelzpunkt: –62 °C
Dichte: 0,981
Flammpunkt: 29 °C (84 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005302
CAS-Nummer: 10431-98-8
EINECS: 233-912-4
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR