2-Methyl-2-oxazolin 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-4,5-dihydrooxazole
159971000
159975000
ACRO159971000EA
85.7
EUR
ACRO159971000
ACRO159975000
2-Methyl-2-oxazolin 99%
2-Methyl-2-oxazolin
Formel:
C₄H₇NO Molecular Weight: 85,11 g/mol Dichte: 1 g/cm³ Flammpunkt: 20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005298 CAS-Nummer: 1120-64-5 EINECS: 214-316-3 UN: 1993 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4330 to 1.4350 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | 1.5 to 2.5 % (MEK) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...