2-Methyl-2-oxazolin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-4,5-dihydrooxazole
B24612.14
B24612.22
B24612.14EA
32.9
EUR
B24612.14
B24612.22
2-Methyl-2-oxazolin ≥99%
2-Methyl-2-oxazolin
Purity: 99%
CAS No.: 1120-64-5
Molecular Formula: C4H7NO
Formula Weight: 85.11
Flash Point: 20°(68°F)
Refractive Index: 1.4340
Boiling Point: 109-111°
Beilstein Reference: 104227
UN No.: 1993
MDL No.: MFCD00005298
Formel:
C₄H₇NO Molecular Weight: 85,11 g/mol Siedepunkt: 109…111 °C Dichte: 1,01 Flammpunkt: 20 °C (68 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005298 CAS-Nummer: 1120-64-5 EINECS: 214-316-3 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...