N-Allylrhodanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Allylrhodanine, 3-Allyl-2-thioxo-4-thiazolidinone

M-Clarity™ Qualitätsstufe = MQ100

A35607-100G A35607-25G
SIALA35607-100GEA 240 EUR
SIALA35607-100G SIALA35607-25G
N-Allylrhodanin, Sigma-Aldrich®
N-Allylrhodanin
Formel: C₆H₇NOS₂
Molecular Weight: 173,26 g/mol
MDL: MFCD00044700
CAS-Nummer: 1457-47-2
EINECS: 215-941-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR