5-(4-(Dimethylamino)benzyliden)rhodanin 98%
Lieferant: Thermo Scientific
Synonyme:
4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone, p-Dimethylaminobenzalrhodanine, 5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-one, p-(Dimethylamino)benzal-5-rhodanine, (5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one
A17152.06
A17152.09
A17152.14
A17152.06EA
38.9
EUR
A17152.06
A17152.09
A17152.14
5-(4-(Dimethylamino)benzyliden)rhodanin 98%
5-(4-(Dimethylamino)benzyliden)rhodanin
Purity: 98%
CAS No.: 536-17-4
Molecular Formula: C12H12N2OS2
Formula Weight: 264.37
Odour: Weak
Melting Point: 275-280°
Beilstein Reference: 189065
Merck Reference: 14,3231
MDL No.: MFCD00064857
Formel:
C₁₂H₁₂N₂OS₂ Molecular Weight: 264,37 g/mol Schmelzpunkt: 275…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064857 CAS-Nummer: 536-17-4 EINECS: 208-625-2 Merck Index: 14,3231 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...