2,1,3-Benzothiadiazol

Lieferant: Thermo Scientific

RDP00044.1 RDP00044.10 RDP00044.50
MAYBRDP00044.1EA 0 EUR
MAYBRDP00044.1 MAYBRDP00044.10 MAYBRDP00044.50
2,1,3-Benzothiadiazol
2,1,3-Benzothiadiazole
Formel: C₆H₄N₂S
Molecular Weight: 136,18 g/mol
MDL: MFCD00005809
CAS-Nummer: 273-13-2
EINECS: 205-985-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR