2,1,3-Benzothiadiazol

Lieferant: COMBI-BLOCKS

HC-3087-25G HC-3087-100G
COBBHC-3087-25GEA 51.3 EUR
COBBHC-3087-25G COBBHC-3087-100G
2,1,3-Benzothiadiazol
2,1,3-Benzothiadiazole
Formel: C₆H₄N₂S
Molecular Weight: 136.18 g/mol
MDL: MFCD00005809
CAS-Nummer: 273-13-2
EINECS: 205-985-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR