2-Methylbenzo[b]thiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

467456-1G 467456-5G
SIAL467456-1GEA 72.7 EUR
SIAL467456-1G SIAL467456-5G
2-Methylbenzo[b]thiophen, Sigma-Aldrich®
2-Methylbenzo[b]thiophen
Formel: C₉H₈S
Molecular Weight: 148,23 g/mol
MDL: MFCD00216250
CAS-Nummer: 1195-14-8
EINECS: 214-792-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR