5-Methylbenzo[b]thiophen
Lieferant: FLUOROCHEM
CFF285.5
CFF285.25
017331-5G
017331-25G
017331-1G
CHMPCFF285.5EA
508
EUR
CHMPCFF285.5
CHMPCFF285.25
FLUO017331-5G
FLUO017331-25G
FLUO017331-1G
5-Methylbenzo[b]thiophen
5-Methylbenzo[b]thiophen
Formel:
C₉H₈S Molecular Weight: 148.23 g/mol |
MDL:
MFCD00052509 CAS-Nummer: 14315-14-1 EINECS: 238-256-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...