3-Methylbenzo[b]thiophen-2-carbaldehyd

Lieferant: FLUOROCHEM

023325-1G 023325-5G
FLUO023325-1GEA 63.9 EUR
FLUO023325-1G FLUO023325-5G
3-Methylbenzo[b]thiophen-2-carbaldehyd
3-Methylbenzo[b]thiophen-2-carbaldehyd
Formel: C₁₀H₈OS
Molecular Weight: 176.24 g/mol
MDL: MFCD00052307
CAS-Nummer: 22053-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR