2-Benzoxazolinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: benzo[d]oxazol-2(3H)-one

M-Clarity™ Qualitätsstufe = MQ200

157058-100G 157058-25G
SIAL157058-100GEA 121 EUR
SIAL157058-100G SIAL157058-25G
2-Benzoxazolinon, Sigma-Aldrich®
2-Benzoxazolinon
Formel: C₇H₅NO₂
Molecular Weight: 135,12 g/mol
MDL: MFCD00005716
CAS-Nummer: 59-49-4
EINECS: 200-430-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR