6,7-Dimethoxy-4-chinazolon
Lieferant: FLUOROCHEM
Synonyme:
6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one
CFF523.1
CFF523.5
CFF523.10
017125-5G
017125-1G
017125-25G
CHMPCFF523.1EA
532
EUR
CHMPCFF523.1
CHMPCFF523.5
CHMPCFF523.10
FLUO017125-5G
FLUO017125-1G
FLUO017125-25G
6,7-Dimethoxy-4-chinazolon
6,7-Dimethoxy-4-chinazolon
Formel:
C₁₀H₁₀N₂O₃ Molecular Weight: 206,2 g/mol |
MDL:
MFCD01570147 CAS-Nummer: 13794-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...