4-Chinazolon ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyquinazoline, Quinazolin-4(3H)-one
A17129.09
A17129.18
A17129.09EA
34.1
EUR
A17129.09
A17129.18
4-Chinazolon ≥98%
4-Chinazolon
Purity: 98%
CAS No.: 491-36-1
Molecular Formula: C8H6N2O
Formula Weight: 146.15
Odour: Odourless
Melting Point: 216-219°
Beilstein Reference: 118473
MDL No.: MFCD00511302
Formel:
C₈H₆N₂O Molecular Weight: 146,15 g/mol Schmelzpunkt: 216…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00511302 CAS-Nummer: 491-36-1 EINECS: 207-735-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...