(1H,3H)-Chinazolin-2,4-dion ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Quinazoline-2,4(1H,3H)-dione, Benzoyleneurea

L07763.06 L07763.14
L07763.06EA 28 EUR
L07763.06 L07763.14
(1H,3H)-Chinazolin-2,4-dion ≥98%
(1H,3H)-Chinazolin-2,4-dion

Purity: 98% CAS No.: 86-96-4 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 300° Beilstein Reference: 383776

Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006699
CAS-Nummer: 86-96-4
EINECS: 201-712-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR