(1H,3H)-Chinazolin-2,4-dion, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Quinazoline-2,4(1H,3H)-dione, Benzoyleneurea
M-Clarity™ Qualitätsstufe = MQ100
142026-100G
142026-25G
SIAL142026-100GEA
172
EUR
SIAL142026-100G
SIAL142026-25G
(1H,3H)-Chinazolin-2,4-dion, Sigma-Aldrich®
(1H,3H)-Chinazolin-2,4-dion
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol |
MDL:
MFCD00006699 CAS-Nummer: 86-96-4 EINECS: 201-712-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...