2,2-Dimethyl-1,3-dioxolan ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L00482.06
L00482.14
L00482.06EA
25.9
EUR
L00482.06
L00482.14
2,2-Dimethyl-1,3-dioxolan ≥98%
2,2-Dimethyl-1,3-dioxolan
Purity: 98+%
CAS No.: 2916-31-6
Molecular Formula: C5H10O2
Formula Weight: 102.13
Flash Point: 6°(42°F)
Refractive Index: 1.3980
Boiling Point: 91-93°
Beilstein Reference: 102775
UN No.: 1993
MDL No.: MFCD00014106
Formel:
C₅H₁₀O₂ Molecular Weight: 102,13 g/mol Siedepunkt: 91…93 °C Dichte: 0,946 Flammpunkt: 6 °C (42 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014106 CAS-Nummer: 2916-31-6 EINECS: 220-842-4 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...