3,4-Dichlor-1,2,5-thiadiazol ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A11738.09 A11738.18 A11738.30
A11738.09EA 34.1 EUR
A11738.09 A11738.18 A11738.30
3,4-Dichlor-1,2,5-thiadiazol ≥99%
3,4-Dichloro-1,2,5-thiadiazole

Purity: 99% CAS No.: 5728-20-1 Molecular Formula: C2Cl2N2S Formula Weight: 155.01 Flash Point: 100°(212°F) Refractive Index: 1.5620 Boiling Point: 157-159° Beilstein Reference: 606585 UN No.: 2810 MDL No.: MFCD00010072

Formel: C₂Cl₂N₂S
Molecular Weight: 155.01 g/mol
Siedepunkt: 157…159 °C
Dichte: 1.644
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010072
CAS-Nummer: 5728-20-1
EINECS: 227-232-7
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR