1,2,3-Thiadiazol-4-carbonsäure

Lieferant: BIONET RESEARCH

9L-544S.500 9L-544S.1 9L-544S.5 9L-544S.10
BIOT9L-544S.500EA 0 EUR
BIOT9L-544S.500 BIOT9L-544S.1 BIOT9L-544S.5 BIOT9L-544S.10
1,2,3-Thiadiazol-4-carbonsäure
1,2,3-Thiadiazole-4-carboxylic acid
Formel: C₃H₂N₂O₂S
MDL: MFCD00052099
CAS-Nummer: 4100-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR