2-Amino-1,3,4-thiadiazol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

L01244.06 L01244.14
L01244.06EA 47.4 EUR
L01244.06 L01244.14
2-Amino-1,3,4-thiadiazol ≥98%
2-Amino-1,3,4-thiadiazole

Purity: 98+% CAS No.: 4005-51-0 Molecular Formula: C2H3N3S Formula Weight: 101.13 Melting Point: 188-194° Beilstein Reference: 107135 UN No.: 2811 MDL No.: MFCD00003107

Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
Schmelzpunkt: 188…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003107
CAS-Nummer: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR