2-Amino-1,3,4-thiadiazol

Lieferant: Thermo Scientific

Synonyme: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

CC08114.250 CC08114.1
MAYBCC08114.250EA 0 EUR
MAYBCC08114.250 MAYBCC08114.1
2-Amino-1,3,4-thiadiazol
2-Amino-1,3,4-thiadiazole
Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
MDL: MFCD00003107
CAS-Nummer: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR