5-Amino-1,2,4-thiadiazol
Lieferant: FLUOROCHEM
Synonyme:
1,2,4-Thiadiazol-5-amine
013903-5G
013903-1G
FLUO013903-5GEA
264
EUR
FLUO013903-5G
FLUO013903-1G
5-Amino-1,2,4-thiadiazol
5-Amino-1,2,4-thiadiazole
Formel:
C₂H₃N₃S |
MDL:
MFCD01102171 CAS-Nummer: 7552-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...