5-Amino-3-methoxy-1,2,4-thiadiazol
Lieferant: FLUOROCHEM
009665-1G
009665-5G
FLUO009665-1GEA
94.1
EUR
FLUO009665-1G
FLUO009665-5G
5-Amino-3-methoxy-1,2,4-thiadiazol
5-Amino-3-methoxy-1,2,4-thiadiazole
Formel:
C₃H₅N₃OS |
MDL:
MFCD03093825 CAS-Nummer: 98022-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...