2-Amino-5-mercapto-1,3,4-thiadiazol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Amino-1,3,4-thiadiazole-2-thiol

A11451.14 A11451.22
A11451.14EA 34.8 EUR
A11451.14 A11451.22
2-Amino-5-mercapto-1,3,4-thiadiazol 98%
2-Amino-5-mercapto-1,3,4-thiadiazole

Purity: 98+% CAS No.: 2349-67-9 Molecular Formula: C2H3N3S2 Formula Weight: 133.19 Melting Point: ca 243° dec. Beilstein Reference: 81728

Formel: C₂H₃N₃S₂
Molecular Weight: 133,2 g/mol
Schmelzpunkt: ∼ 243 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003108
CAS-Nummer: 2349-67-9
EINECS: 219-078-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR