2-Amino-5-mercapto-1,3,4-thiadiazol

Lieferant: CHEMPUR

Synonyme: 5-Amino-1,3,4-thiadiazole-2-thiol

SB00792.250 SB00792.1
CHMPSB00792.250EA 0 EUR
CHMPSB00792.250 MAYBSB00792.1
2-Amino-5-mercapto-1,3,4-thiadiazol
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel: C₂H₃N₃S₂
Molecular Weight: 133,2 g/mol
MDL: MFCD00003108
CAS-Nummer: 2349-67-9
EINECS: 219-078-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR