2,5-Dimercapto-1,3,4-thiadiazol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,3,4-Thiadiazole-2,5-dithiol
115310250
115311000
115310050
ACRO115310250EA
35
EUR
ACRO115310250
ACRO115311000
ACRO115310050
2,5-Dimercapto-1,3,4-thiadiazol 98%
2,5-Dimercapto-1,3,4-thiadiazole
Formel:
C₂H₂N₂S₃ Molecular Weight: 150,25 g/mol Schmelzpunkt: 161…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003103 CAS-Nummer: 1072-71-5 EINECS: 214-014-1 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 156 to 162 °C (Dec) |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...