2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

004036-5G 004036-1G
FLUO004036-5GEA 74.1 EUR
FLUO004036-5G FLUO004036-1G
2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formel: C₃H₂F₃N₃S
Molecular Weight: 169,13 g/mol
MDL: MFCD00003109
CAS-Nummer: 10444-89-0
EINECS: 233-930-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR