2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

M-Clarity™ Qualitätsstufe = MQ200

196967-25G 196967-5G 196967-1G
SIAL196967-25GEA 203 EUR
SIAL196967-25G SIAL196967-5G SIAL196967-1G
2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol, Sigma-Aldrich®
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formel: C₃H₂F₃N₃S
Molecular Weight: 169,13 g/mol
MDL: MFCD00003109
CAS-Nummer: 10444-89-0
EINECS: 233-930-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR