2-Amino-5-ethyl-1,3,4-thiadiazol

Lieferant: CHEMPUR

Synonyme: 5-Ethyl-1,3,4-thiadiazol-2-amine

FL13403.POR 013403-25G
CHMPFL13403.POREA 0 EUR
CHMPFL13403.POR FLUO013403-25G
2-Amino-5-ethyl-1,3,4-thiadiazol
2-Amino-5-ethyl-1,3,4-thiadiazole
Formel: C₄H₇N₃S
Molecular Weight: 129,19 g/mol
MDL: MFCD00003111
CAS-Nummer: 14068-53-2
EINECS: 237-921-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR