2-Amino-5-ethyl-1,3,4-thiadiazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Ethyl-1,3,4-thiadiazol-2-amine
M-Clarity™ Qualitätsstufe = MQ200
196924-25G
196924-5G
SIAL196924-25GEA
251
EUR
SIAL196924-25G
SIAL196924-5G
2-Amino-5-ethyl-1,3,4-thiadiazol, Sigma-Aldrich®
2-Amino-5-ethyl-1,3,4-thiadiazole
Formel:
C₄H₇N₃S Molecular Weight: 129,19 g/mol |
MDL:
MFCD00003111 CAS-Nummer: 14068-53-2 EINECS: 237-921-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...