2-Amino-5-methyl-1,3,4-thiadiazol
Lieferant: COMBI-BLOCKS
OS-7675-5G
OS-7675-25G
OS-7675-100G
COBBOS-7675-5GEA
26.2
EUR
COBBOS-7675-5G
COBBOS-7675-25G
COBBOS-7675-100G
2-Amino-5-methyl-1,3,4-thiadiazol
2-Amino-5-methyl-1,3,4-thiadiazole
Formel:
C₃H₅N₃S Molecular Weight: 115,16 g/mol |
MDL:
MFCD00003110 CAS-Nummer: 108-33-8 EINECS: 203-573-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...