2-Amino-5-methyl-1,3,4-thiadiazol

Lieferant: CHEMPUR

FL9899.POR 009899-1G 009899-5G
CHMPFL9899.POREA 0 EUR
CHMPFL9899.POR FLUO009899-1G FLUO009899-5G
2-Amino-5-methyl-1,3,4-thiadiazol
2-Amino-5-methyl-1,3,4-thiadiazole
Formel: C₃H₅N₃S
Molecular Weight: 115,16 g/mol
MDL: MFCD00003110
CAS-Nummer: 108-33-8
EINECS: 203-573-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR