5-Amino-3-isopropyl-1,2,4-thiadiazol
Lieferant: FLUOROCHEM
009664-1G
009664-5G
FLUO009664-1GEA
110
EUR
FLUO009664-1G
FLUO009664-5G
5-Amino-3-isopropyl-1,2,4-thiadiazol
5-Amino-3-isopropyl-1,2,4-thiadiazol
Formel:
C₅H₉N₃S |
MDL:
MFCD03093824 CAS-Nummer: 32039-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...