5-Amino-3-ethyl-1,2,4-thiadiazol
Lieferant: FLUOROCHEM
009733-1G
009733-5G
FLUO009733-1GEA
110
EUR
FLUO009733-1G
FLUO009733-5G
5-Amino-3-ethyl-1,2,4-thiadiazol
5-Amino-3-ethyl-1,2,4-thiadiazole
Formel:
C₄H₇N₃S |
CAS-Nummer:
17467-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...